Skip to Content
Merck
All Photos(1)

Key Documents

CLS3783

Corning® Transwell® HTS receiver plate

size 96 wells, white plate, sterile

Synonym(s):

96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, EIA/RIA plate, immunoassay plate

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$259.00

$259.00


Check Cart for Availability


Select a Size

Change View
10 EA
$259.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
flat wells
white bottom
white plate
white wells
size:
96 wells

$259.00


Check Cart for Availability

material

flat wells
white bottom
white plate
white wells

description

an accessory for the HTS Transwell-96 Permeable Support system

sterility

sterile

feature

lid
skirt

packaging

pack of 10 ea

manufacturer/tradename

Corning 3783

size

96 wells

color

white

suitability

suitable for (cell cuture)

binding type

Tissue Culture (TC)-treated surface

General description

  • Plate is treated for optimal cell attachment
  • For use in cell growth or assay steps of an experiment
  • Used for luminescent evaluation
  • When used with HTS Transwell®-96 unit, the plate allows for convenient pipet access to both apical and basolateral sides of each well (manual or automated)
  • Lids included
  • TC surface treated

Legal Information

Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service