Skip to Content
Merck
All Photos(1)

Key Documents

CLS3686

Corning® 96 Well Half-Area Microplate

flat bottom, black polystyrene, NBS (non-binding surface), bag of 25 inner package, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$1,200.00

$1,200.00


Check Cart for Availability


Select a Size

Change View
100 EA
$1,200.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
black bottom
black polystyrene
flat bottom
size:
96 wells

$1,200.00


Check Cart for Availability

material

black bottom
black polystyrene
flat bottom

sterility

non-sterile

feature

lid: no
skirt

packaging

bag of 25 inner package

manufacturer/tradename

Corning 3686

size

96 wells

well D

10.7 mm

well volume

190 μL

well working volume

25-125 μL

color

black

suitability

suitable for (cell cuture)

binding type

NBS (non-binding surface)

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Features and Benefits

  • Corning® 96 well half-area microplates support various application requirements.
  • Black microplates reduces background and light scattering, resulting in higher signal-to-noise ratios.
  • Designed to reduce well-to-well crosstalk.
  • Non-treated polystyrene surface is suitable primarily for the immobilization of large molecules, such as antibodies.
  • The flat bottom is usually the best choice for detection.

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service