Skip to Content
Merck
All Photos(1)

Key Documents

CLS3601

Corning®  96 Well Black Polystyrene Microplate

flat bottom wells clear, black polystyrene, high binding surface, bag of 25, non-sterile, lid: no

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$969.00

$969.00


Available to ship on14 April 2025Details



Select a Size

Change View
100 EA
$969.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
black polystyrene
flat bottom wells clear
polystyrene
round clear wells
size:
96 wells

$969.00


Available to ship on14 April 2025Details


material

black polystyrene
flat bottom wells clear
polystyrene
round clear wells

sterility

non-sterile

feature

lid: no
skirt
plate format: 96 well standard

packaging

case of 100
bag of 25

manufacturer/tradename

Corning 3601

size

96 wells

well volume

360 μL

well working volume

75-200 μL

color

black

suitability

suitable for (fluorescent/luminescent assays)

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

  • High binding surface binds medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions
  • Binding capacity is approximately 400 to 500 ng IgG/cm2
  • Ideal for immunoassays
  • Without lids, lids supplied separately
  • Bottoms are 60% thinner than conventional polystyrene plates, resulting in lower background fluorescent and enabling readings down to 340 nm
  • Designed to reduce well-to-well crosstalk during fluorescent assays
  • Can be used for both top and bottom reading instruments
  • Recommended working volume of 75 to 200 μL
  • Conforms to ANSI/SBS standard microplate footprint and dimensions

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service