Skip to Content
Merck
All Photos(2)

Key Documents

CLS356521

Corning® Falcon® Serological Pipettes

capacity 1 mL, individually wrapped, clear-plastic, sterile

Synonym(s):

pipet

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.02

material

polystyrene

sterility

sterile

feature

individually wrapped clear-plastic
subdivision

manufacturer/tradename

Corning 356521

capacity

1 mL

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Liquid Handling

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service