CLS351007
Corning® Falcon® Bacteriological Petri Dish
W × H 60 mm × 15 mm, lid, standard, sterile
Synonym(s):
plastic cell culture dish
About This Item
Recommended Products
material
polystyrene
sterility
sterile
feature
lid standard
manufacturer/tradename
Corning 351007
technique(s)
microbiological culture: suitable
W × H
60 mm × 15 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service