Key Documents
CLS3081200LO
Pyrex® large knob top desiccators
replacement desiccator bowl for 5.8L large knob top desiccator, 1/cs
Synonym(s):
desiccator bowls, desiccator lower bowls
Select a Size
1 EA
$657.00
$657.00
Check Cart for Availability
Select a Size
Change View
1 EA
$657.00
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
$657.00
Check Cart for Availability
material
borosilicate glass
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 3081-200LO
suitability
suitable for (general analytical work)
Related Categories
General description
Note: This is the replacement desiccator bowl for the Pyrex desiccator CLS3181200.
The Pyrex large knob top desiccators are designed for general analytical work. The large sturdy knob makes the cover easy to handle. Warning: Not for use with vacuum applications. Do not subject to heat or pressure.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)