Key Documents
CLS3081150CO
Pyrex® small knob top desiccator
cover only
Synonym(s):
equipment replacement accessory
Select a Size
1 EA
$202.00
$202.00
Check Cart for Availability
Select a Size
Change View
1 EA
$202.00
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
$202.00
Check Cart for Availability
material
borosilicate glass
manufacturer/tradename
Corning 3081-150CO
Related Categories
General description
This 2.4 L Pyrex desiccator is designed for general analytical work. The sturdy knob makes the cover easy to handle.
Warning: Do not heat or subject to pressure. Not for vacuum applications.
This is the desiccator cover replacement which fits Pyrex desiccator CLS3181150.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)