CLS2975243
Corning® cover glasses
rectangular, No. 1, W × L 24 mm × 30 mm
Synonym(s):
coverslips, glass covers, slide covers
Select a Size
$36.40
Select a Size
About This Item
$36.40
Recommended Products
material
borosilicate glass
rectangular
packaging
pack of 100 ea
case of 1000 ea
manufacturer/tradename
Corning 2975-243
W × L
24 mm × 30 mm
binding type
non-treated surface
Features and Benefits
- Corning® cover glasses are made from optically clear borosilicate glass.
- Chemically resistant cover glasses are not affected by weathering or surface attachment.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)