CLS2484
Corning® 1 x 8 Stripwell™ 96 well plates
low volume, medium binding surface, bulk packed, white
Synonym(s):
1 x 8 strip plate
Select a Size
$1,240.00
Select a Size
About This Item
round white wells
white polystyrene plate
$1,240.00
Recommended Products
material
flat bottom clear polystyrene (96-well)
round white wells
white polystyrene plate
sterility
non-sterile
feature
lid: no
packaging
plate of 12 strips
manufacturer/tradename
Corning 2484
size
96 wells
well volume
190 μL
well working volume
75-200 μL , for low volume plates volume is 50-125 μL
color
white
suitability
suitable for (cell cuture)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service