CLS11465
Corning® Erlenmeyer flask, with aseptic connector
volume 2 L, I.D. 1/4 in., sterile
Synonym(s):
Corning container
Select a Size
$503.00
Select a Size
About This Item
$503.00
Recommended Products
material
polycarbonate
description
plain
sterility
sterile
feature
cap: no
C-Flex tubing
packaging
case of 2
manufacturer/tradename
Corning 11465
I.D.
1/4 in.
capacity
2 L
volume
2 L
20000 mL
fitting
male luer lock
Looking for similar products? Visit Product Comparison Guide
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service