Skip to Content
Merck
All Photos(1)

Key Documents

CLLS1003

Sigma-Aldrich

DLD1 Cells PIK3CA +/-

human male colon (Source Disease: Colorectal adenocarcinoma)

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$5,660.00

$5,660.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 SET
$5,660.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

$5,660.00


Check Cart for Availability

Request a Bulk Order

Product Name

DLD1 Cells PIK3CA +/-,

biological source

human male colon (Source Disease: Colorectal adenocarcinoma)

Quality Level

OMIM accession no.

storage temp.

−196°C

Gene Information

human ... PIK3CA(5290)

General description

DLD1 Cells PIK3CA +/- are colorectal adenocarcinoma, epithelial cells, from a human adult male, with a ZFN knock out modification.

Application

Media Renewal changes two to three times per week
Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.
Subculture Ratio: approx. 1:3 to 1:10
The base medium for this cell line is RPMI, Cat. No. R5886. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration (v/v) of 10%.
The cell freezing medium is DMSO 1X (Cat. No. C6164)

Features and Benefits

These DLD-1 cells are adherent, with a doubling time of approx. 20 hours.
Zinc finger nuclease (ZFN) knock out on chromosome 3q23.3

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, PCR assay for cell line species confirmation.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Kit Components Also Available Separately

Product No.
Description
SDS

  • DLD1 Control Cells 1 vial

  • DLD1 Cells PIK3CA +/- 1 vial

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service