Skip to Content
Merck
All Photos(1)

Key Documents

CLL1221

Sigma-Aldrich

Safe Harbor Landing Pad Cell Line Jurkat T-Lymphocytes

human male peripheral blood (Source Disease: Acute T cell leukemia)

Synonym(s):

T-Lymphocyte Cell Line

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$6,140.00

$6,140.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
$6,140.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

$6,140.00


Check Cart for Availability

Request a Bulk Order

Product Name

Safe Harbor Landing Pad Cell Line Jurkat T-Lymphocytes,

biological source

human male peripheral blood (Source Disease: Acute T cell leukemia)

Quality Level

form

frozen liquid (Vial of Frozen Cells)

growth mode

Suspension

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

The STR profile of this cell line matches that of its parental cell line ATCC® Catalog No. TIB-152. Jurkat T lymphocytes are a human, acute T cell lymphoma cell line isolated in the late 1970s from the peripheral blood of a young male patient suffering from T cell leukemia. The cells possess a pseudodiploid karyotype and have been characterized as expressing CD3 and, upon stimulation, interleukin-2.

Cell Line Description

These cells are a human, acute T cell lymphoma cell line isolated from the peripheral blood of a young male suffering from T cell leukemia. The cells possess a pseudodiploid karyotype, express CD3 and, upon stimulation, IL-2.

Application

This product is a human Jurkat T-lymphocyte cell line in which a landing pad cassette has been integrated into the AAVS1 safe harbor locus using CompoZr® Zinc Finger Nuclease technology. The mKATE2 fluorescence gene was integrated following the EF1a promoter and flanked by unique Cre-lox sites. The design of this landing pad cassette allows for easy, fast, and affordable genetic modification using Cre recombinase. mKATE2 can easily be exchanged for a payload of the user′s choice using Cre recombinase and a targeting vector with appropriate lox sites. Cells can then be sorted via fluorescence-activated cell sorting (FACS) for loss of mKATE2 expression as a surrogate for successful integration of the targeting vector. Approximately 7-10 days are required for loss of the mKATE2 signal in successfully targeted cells. See technical bulletin for detailed protocols.

Features and Benefits

These cells contain the mKATE2 fluorescence gene flanked by unique Cre-lox sites inserted in the AAVS1 safe harbor gene. These Jurkat cells are grown in suspension with a doubling time of approximately 22 hours.

Quality

Tested for Mycoplasma, bacterial and fungal content, post-freeze viability, short terminal repeat (STR) analysis for cell line identification.

Culture Medium

RPMI modified to contain 2mM L-glutamine, 10mM HEPES, 1mM sodium pyruvate, and 1500mg/L sodium bicarbonate. Add 10% FBS(Catalog Number F2442).

Legal Information

ATCC is a registered trademark of American Type Culture Collection
CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huseyin Tas et al.
PloS one, 10(9), e0136963-e0136963 (2015-09-04)
We describe an optimized system for the easy, effective, and precise modification of the Escherichia coli genome. Genome changes are introduced first through the integration of a 1.3 kbp Landing Pad consisting of a gene conferring resistance to tetracycline (tetA)
Zhong-Wei Du et al.
Stem cells (Dayton, Ohio), 27(5), 1032-1041 (2009-05-06)
To circumvent the silencing effect of transgene expression in human embryonic stem cells (hESCs), we employed the Cre recombination-mediated cassette exchange strategy to target the silencing-resistant site in the genome. We have identified new loci that sustain transgene expression during
Kimi Araki et al.
Nucleic acids research, 30(19), e103-e103 (2002-10-05)
The Cre-lox system is an important tool for genetic manipulation. To promote integrative reactions, two strategies using mutant lox sites have been developed. One is the left element/right element (LE/RE)-mutant strategy and the other is the cassette exchange strategy using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service