Skip to Content
Merck
All Photos(1)

Documents

MFM-223

98050130, human breast, Epithelial-like

Synonym(s):

MFM223 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MFM-223, 98050130

biological source

human breast

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Not specified

receptors

Androgen receptor

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human caucasian mammary carcinoma

Cell Line Description

The mammary carcinoma cell line MFM-223 was obtained from the pleural effusion of a postmenopausal breast cancer patient. The patient had not received any prior treatment and the tumour was classified as a widespread ductal mammary carcinoma with the histological grade of III. The cell line is characterised by high androgen, (approximately 100,000 binding sites per cell) and low oestrogen and progesterone receptor levels. Proliferation of cells was inhibited by addition of the androgen dihydrotestosterone at concentrations above 0.01 nM. MFM-223 have a characteristic pattern of isoenzyme polymorphism and of karyotype alterations revealing marker chromosomes and homogenously staining regions. Most cells have been near diploid with 10% displaying near tetraploidy. The cells have been tested to be free of Hepatitis B and C viruses and HIV. MFM-223 is tumourigenic in nude mice.

Application

Androgen receptor studies, study of mammary carcinoma

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 8,12
D16S539: 11
D5S818: 11,12
D7S820: 8,10
THO1: 6,9.3
TPOX: 8
vWA: 17

Culture Medium

MEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service