EMT6/VRP/R
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
mouse unknown/unspecified
packaging
tube of 5 μg 96042319-DNA-5UG
pkg of vial of cells 96042319-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse mammary, drug-resistant
Cell Line Description
The drug-resistant line EMT6/VRP/R was established from the parent cell line EMT6/P (Sigma Catalogue number. 96042344) by growth in verapamil (see reference).
Application
Drug-resistance studies
Culture Medium
MEM (EBSS) +2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 75 μg/ml verpamil + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 using 0.25% trypsin or trypsin/EDTA; 5% CO2;37ºC. It is recommended to resuscitate cells in drug-free medium. Add drug after growth is established.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service