COR-L32
96020744, human unknown/unspecified, Not specified
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
Product Name
COR-L32, 96020744
biological source
human unknown/unspecified
growth mode
Aggregates in suspension
karyotype
Not specified
morphology
Not specified
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human small cell carcinoma
Cell Line Description
The cell line COR-L32 has been estalished from a marrow trephine from a poorly-differentiated squamous cell carcinoma. The patient has been confirmed to have primary lung cancer. Electron micrographs showed dense core granules and other features of neuroendocrine differentiation. In culture, COR-L32 grow as floating aggregates of packed cells.
Application
Lung cancer study
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Maintain cultures between 3-9 x 100,000 cells / ml; 5% CO2; 37°C. This is a human small cell carcinoma which grows in large clumps. Cell banks should be frozen at high cell densities e.g. 6-9 x 1000,000 cells/ml to allow for clumping.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service