Skip to Content
Merck
All Photos(1)

Key Documents

XrS6-hamKu80

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

hamster ovary

packaging

tube of 5 μg 96012301-DNA-5UG
pkg of vial of cells 96012301-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Ku80

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Hamster Chinese ovary, transformed with hamster Ku80 cDNA

Cell Line Description

The cell line Xrs6-hamKu80 was derived from a radiosensitive CHO-K1 mutant transfected with hamster Ku80 cDNA. Ku80, a subunit of the Ku protein and component of the DNA-dependent protein kinase, is a gene product defective in radiosensitive CHO cells, so called xrs mutants. XRC5, the gene encoding Ku80, belongs to the ionisation radiation group affected in DNA repair defects. Mutants transfected with hamster Ku80 obtained full restoration of gamma-ray resistance, DNA end-binding activity to parent levels and expression of Ku80 and Ku70 proteins.

Application

Study of DNA repair mechanisms

Culture Medium

Ham′s F12 + 2mM Glutamine + 300μg/ml G418 + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service