Skip to Content
Merck
All Photos(1)

Documents

NCI-H358

95111733, human lung (bronchoalveolar), Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NCI-H358, 95111733

biological source

human lung (bronchoalveolar)

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Caucasian bronchioalveolar carcinoma

Cell Line Description

NCI-H358 was isolated from a primary bronchioalveolar carcinoma of the lung from a Caucasian male taken prior to treatment. Ultrastructural studies of this non-small cell carcinoma of the lung (NSCLC) demonstrated the presence of granules characteristic of Clara cells. NCI-H358 do not express UDP-glucuronosyltransferases, but do express glutathione-S-transferase and phenol sulphotransferase. Expression of SP-A protein and RNA, the major surfactant-associated protein was detected. SP-B and SP-C RNA was not expressed. A complete homozygous deletion of the p53 gene and therefore a lack of p53 protein has been reported. A colony forming efficiency of 0.83% in soft agarose, and growth in serum-free media has been reported. The cells are tumourigenic in athymic nude mice, and exhibit a doubling time of 38 hours in RPMI 1640 medium. Since these cells are proficient in oxidation of xenobiotics but deficient in their conjugation with glucuronic acid they present a tool for analysing the role of glucuronic acid conjugation in the inactivation of chemicals in intact cells.

Application

Test system for evaluating cytotoxicity and genotoxicity of chemicals to human lung

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 8,12
D16S539: 12,13
D5S818: 10,12
D7S820: 10,11
THO1: 6
TPOX: 8,9
vWA: 17

Culture Medium

RPMI 1640 + 2mM Glutamine + 5-10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service