AFRC 84B4
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
Horse x Mouse/Horse Hybridoma
packaging
tube of 5 μg 94120829-DNA-5UG
pkg of vial of cells 94120829-1VL
ImmunoDon
Horse sub-mandibular lymph node
Myeloma
AFRC 17D1
Reactivity
Anti-Haemagglutinin, influenza
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Horse sub-mandibular lymph node x AFRC 17D1
Cell Line Description
The heterohybridoma AFRC 84B4 is one of 8 hybridoma cell lines obtained using lymphocytes from ponies immunised with influenza A equine 2 virus (isolateA/Equine/Newmarket/79 (H3N8)). For the establishment of AFRC 84B4 horse cells from the sub-mandibular lymph node have been fused with the non-Ig-secreting heterohybridoma AFRC 17D1 (Sigma Catalogue number. 94120825).
Immunogen
Influenza A equine 2 virus
Application
Haemagglutinin detection
The isotype of the monoclonal antibody produced by this cell line is horse IgG.
Culture Medium
RPMI 1640 + 2mM glutamine + 1mM Sodium Pyruvate (NaP) + 5% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service