Skip to Content
Merck
All Photos(1)

Documents

FTC-238

94060902, human thyroid (lung metastasis), Polymorphic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

FTC-238, 94060902

biological source

human thyroid (lung metastasis)

growth mode

Adherent

karyotype

Not specified

morphology

Polymorphic

products

Expression of 5′ - Deiodinase Type I

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human follicular thyroid carcinoma, lung metastasis

Cell Line Description

FTC-238 was established from a lung metastasis of a follicular thyroid carcinoma from a 42-year-old male. The cells are polymorphic showing flat polygonal to spindle-like morphologies. Complex chromosomal changes were detected as well as a p53 mutation. FTC-133 (Sigma Catalogue number 94060901), FTC-236 (Sigma Catalogue number 06030202) and FTC-238 (Sigma Catalogue number 94060902) were all derived from the same individual, a 42 year old male with follicular thyroid cancer. This has been confirmed by short tandem repeat STR-PCR analysis at ECACC The Y chromosome could not be detected in these cell line by (STR)-PCR analysis. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell lines are identical to the source provided by the depositor based on the STR-PCR analysis.

Application

FTC-238 has been used to study the function of human thyroid and development of thyroid cancer.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,11
D13S317: 11
D16S539: 11,12
D5S818: 11,12
D7S820: 9,10
THO1: 9.3
TPOX: 9
vWA: 15,18

Culture Medium

DMEM:Ham′s F12 (1:1) + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 to 1:20 i.e. seeding at 1-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service