CALU 1
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 93120818, human lung, Epithelial
Synonym(s):
Calu1 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
human lung
packaging
tube of 5 μg 93120818-DNA-5UG
pkg of vial of cells 93120818-1VL
growth mode
Adherent
karyotype
Hypotriploid, modal no. 62. No Y chromosome.
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian lung epidermoid carcinoma
Cell Line Description
Derived from a 47 year old Caucasian male with an epidermoid carcinoma of the lung (grade III). The line will form epidermoid carcinomas in nude mice. Tumours also develop in steroid treated hamsters. Ultrastructure consists of numerous microvilli, prominent rough endoplasmic reticulum, lysosomes, lipid inclusions and no virus particles. The HLA profile is: A-10,11 B15, w35. The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.
Application
Tumourigenicity studies
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 11,12
D16S539: 11
D5S818: 10,12
D7S820: 9,10
THO1: 9.3
TPOX: 8
vWA: 15,16
CSF1PO: 10
D13S317: 11,12
D16S539: 11
D5S818: 10,12
D7S820: 9,10
THO1: 9.3
TPOX: 8
vWA: 15,16
Culture Medium
EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service