Skip to Content
Merck
All Photos(1)

Documents

BXPC3 Cell Line human

93120816, human pancreas, Epithelial

Synonym(s):

Biopsy Xenograft of Pancreatic Carcinoma Cell Line-3, BxPC-3 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

BXPC3 Cell Line human, 93120816

biological source

human pancreas

description

Human primary pancreatic adenocarcinoma

growth mode

Adherent

karyotype

2n = 59, near triploid

morphology

Epithelial

products

Mucin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human primary pancreatic adenocarcinoma

Cell Line Description

Derived from a 61 year old female with a primary adenocarcinoma of the pancreas. BxPC-3 cells produce mucin and the tumour produced in a nude mouse is moderately well to poorly differentiated like the primary adenocarcinoma.

Application

BXPC3 cell line has been used to determine the biological and therapeutic importance of gastrin gene expression in pancreatic adenocarcinomas.
Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 13
D13S317: 11
D16S539: 9,11
D5S818: 11
D7S820: 10,13
THO1: 9
TPOX: 8
vWA: 14,18

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

ECACC pancreatic cancer cell lines for various applications including drug response studies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service