Skip to Content
Merck
All Photos(1)

Key Documents

BE10-Late

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

rat brain

packaging

tube of 5 μg 93040120-DNA-5UG
pkg of vial of cells 93040120-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Plasminogen activator activity

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat, BDIX, foetal brain, malignant

Cell Line Description

Malignant cells produced on passaging BE10-Intermediate (Sigma Catalogue number. 93040119) beyond passage 40. The cells possess tissue plasminogen activator activity, are tumourigenic in BDIX rats and form colonies in agar.

Application

Study of carcinogenesis, tumourigenicity studies

Culture Medium

DMEM + 2mM Glutamine + 15% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 to 1:20 i.e. seeding at 1-2x100,000 cells/25cm2 using 0.25% trypsin/EDTA; 10-15% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service