Skip to Content
Merck
All Photos(1)

Documents

U28

Reactivity: Anti-IgG2a, mouse, 93022518

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0

Reactivity

Anti-IgG2a, mouse

technique(s)

cell culture | mammalian: suitable

isotype

IgM

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Mouse BALB/c spleen

Cell Line Description

The hybridoma U28 was obtained by fusing SP2/0 myeloma cells with spleen cells from BALB/c mice that had been given repeated intranasal infection with a single i.v. injection before fusion. The antibody binds murine IgG2a of "a" allotype and the Fc region. There is no cross reactivity with A/PR/8/34 influenza virus, DNP, NP or DNA cytoskeletal proteins. Contains a CH2 domain.

Immunogen

A/PR/8/34 mouse pathogenic human influenza virus

Application

Differential detection of complexed IgG2a in the presence of native IgG2a, allotype detection or quantification

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service