Skip to Content
Merck
All Photos(1)

Documents

AK-D

89071903, feline lung, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

AK-D, 89071903

biological source

feline lung

growth mode

Adherent

karyotype

Diploid

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Foetal cat lung normal

Cell Line Description

A clonal derivative of mass cultures of foetal feline lung after 14 passages in vitro. Susceptible to panleukopenia, feline herpes and feline picornaviruses.

Application

Virus studies: panleucopenia, feline herpes and feline picornaviruses

Culture Medium

Ham′s F12 + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:5 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin and 0.03% EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service