Skip to Content
Merck
All Photos(1)

Documents

P22

89030317, hamster ovary, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

P22, 89030317

biological source

hamster ovary

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Hamster Chinese ovary

Cell Line Description

Chinese hamster ovary cell line which lacks ornithine decarboxylase activity.

Culture Medium

EMEM (EBSS) + Ham′s F12 (1:1) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 0.1% BSA.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 5x1,000 to 1x10,000 cells/cm2 using 0.1% cold PBS and 0.25% cold trypsin. Place flasks at 4°C for 5-10 min. or until cells detach; 5% CO2; 37°C. Pre-treat culture flasks with porcine gela

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service