Product Name
CCRF-HSB-2, 85112801, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
biological source
human blood
packaging
tube of 5 μg 85112801-DNA-5UG pkg of vial of cells 85112801-1VL
A T lymphoblastoid line obtained from the peripheral blood of an 11 year old Caucasian male with acute lymphoblastoid leukaemia which has been passaged eight times through new-born Syrian Hamsters. These may be grown to a high density in a spinner type suspension culture.
Maintain cultures between 5-8x100,000 cells/ml; 5% CO2; 37°C. Maximum cell density 2-3x1,000,000 cells/ml.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)