Skip to Content
Merck
All Photos(1)

Key Documents

SPC111

11120716, human lung (effusion), Epitheloid/mesenchymal

Synonym(s):

SPC-111, SPC111

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

SPC111, 11120716

biological source

human lung (effusion)

growth mode

Adherent

karyotype

Not specified

morphology

Epitheloid/mesenchymal

receptors

Mesothelial-membrane antigen ME1

relevant disease(s)

cancer

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

A human mesothelioma cell line derived from a malignant mesothelioma. Asbestos exposure

Cell Line Description

SPC111 was derived from the pleural effusion of a 55-year old male patient, prior to treatment, with a known history of exposure to asbestos.

DNA Profile

STR-PCR Data:

Amelogenin: X,Y
CSF1PO: 12
D13S317: 12
D16S539: 11,13
D5S818: 10,13
D7S820: 8,10
THO1: 9.3
TPOX: 9
vWA: 18

Culture Medium

DMEM:F12 (1:1) (D8062) + L-Glutamine (G7513) + 15% FBS / FCS (F2442).

Subculture Routine

Split subconfluent cultures (70-80%) 1:4 to 1:10 using 0.25% trypsin/EDTA; 5% CO2; 37 °C. Suggested seeding density 1-4 x 10,000 cells/cm2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service