Skip to Content
Merck
All Photos(1)

Documents

KARPAS 1718

08072401, human spleen (lymphoma), Single round to polymorphic cells growing in suspension

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

KARPAS 1718, 08072401

biological source

human spleen (lymphoma)

growth mode

Suspension

karyotype

Not specified

morphology

Single round to polymorphic cells growing in suspension

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human splenic lymphoma

Cell Line Description

KARPAS 1718 is derived from the peripheral blood of a patient diagnosed with transformed splenic lymphoma with villous lymphocytes

Application

Oncology research

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,13
D13S317: 8,13
D16S539: 11,13
D5S818: 11,12
D7S820: 8
THO1: 6
TPOX: 8,11
vWA: 14

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9 x 105 cells/ml; 5% CO2; 37°C. Thaw ampoule and perform a cell count, spin out the DMSO and resuspend in an appropriate volume of media to give a cell concentration of 4-5 x 105 cells/ml. Viability may be poor on resuscitation and may decrease further within the first 48 hours, the culture may appear non-viable at this time. Clusters of cells will eventually appear but this could take 1-2 weeks, during this time large media changes are not recommended. It may take 2-3 weeks for vigorous growth to be established. At subculture perform a cell count and maintain cultures between 3-9 x 105 cells/ml, 5% CO2, 37°c. For cryopreservation of KARPAS 1718, freeze at approximately 2 x 107 cells/ml in 90% FBS + 10% DMSO.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service