Skip to Content
Merck
All Photos(1)

Documents

54SCC3

8062561, Mouse x Rat Hybridoma nerve, oligopolar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

54SCC3, 08062561

biological source

Mouse x Rat Hybridoma nerve

growth mode

Adherent

karyotype

Modal No. varied with passage see reference

morphology

oligopolar

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat glioma x mouse L cell fibroblast hybrid

Cell Line Description

Hybrid generated by fusion of bromodeoxyuridine-resistant mutant of rat glioma cell line C6 (C6-BU-1, Sigma Catalogue number 08062563) and the 6-thioguanine-resistant mouse L cell fibroblast line A9 (Sigma Catalogue number 08062526) using inactivated Sendai virus. Cells formed from such a fusion are multipolar and extend small processes.

Culture Medium

DMEM + 2 mM glutamine + 10% Foetal Bovine Serum (FBS) + HAT (0.1 mM hypoxanthine, 10 μM aminopterin and 16 μM thymidine) Cells can be cultured without HAT provided they were cultured in between in HT medium for 3-4 days

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service