Skip to Content
Merck
All Photos(1)

Documents

XCIII-3D8-D8

Reactivity: Anti-Leishmania (L) pifanoi -Amastigote, 08041724

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

Balb/c mouse spleen

Myeloma

NS-1 (P3-X63/Ag8-NS1)

Reactivity

Anti-Leishmania (L) pifanoi -Amastigote

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse NS1 x BALB/c mouse

Cell Line Description

XCIII-3D8-D8 was developed by immunizing mice with membranes from Leishmania (L) pifanoi and fusing the mouse spleen cells with NS-1(P3-X63/Ag8-NS1)

Immunogen

Membrane from Leishmania (L) pifanoi strain LTRod-MHOM/VE/60/LtRod-Cl-A-3

Application

Detection of Leishmania (L) pifanoi

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS

Subculture Routine

Seed cultures between 1-3x100,000 cells/ml; Saturation density 1,000,000 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service