Skip to Content
Merck
All Photos(1)

Key Documents

HCA-2

6061901, human colon, Epithelial

Synonym(s):

HCA 2, HCA2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human colon
growth mode:
Adherent
karyotype:
Not specified
morphology:
Epithelial
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer
Pricing and availability is not currently available.

Product Name

HCA-2, from human colon, 06061901

biological source

human colon

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Cell Line Origin

Human sigmoid colon adenocarcinoma

Cell Line Description

Established from a sigmoid colon adenocarcinoma of a 83 year-old female patient. The adenocarcinoma was well differentiated and determined to be Dukes′ stage C. Imperial College Innovations has stated that this cell line is not to be supplied to commercial organisations.

DNA Profile

Not specified

Culture Medium

DMEM + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS)

Subculture Routine

Split subconfluent cultures (70-80%) 1:2 to 1:10 using 0.05% trypsin/EDTA; 7.5% CO2; 37 °C. Suggested seeding density 2-4x10,000 cells/cm2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service