Skip to Content
Merck
All Photos(1)

Key Documents

CAS9RFPV

Sigma-Aldrich

Cas9 RFP Lentiviral Particles

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$2,150.00

$2,150.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$2,150.00

About This Item

UNSPSC Code:
41106621
NACRES:
NA.51

$2,150.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 8 x 25 μL (aliquots)

concentration

≥5 x 10 8 VP/ml virus (via p24 assay)

application(s)

CRISPR

reporter gene

RFP

shipped in

dry ice

storage temp.

−20°C

General description

Ready-to-use Cas9-RFP lentiviral particles enable immediate transduction of a wide range of cell lines. Cas9-RFP lentiviral particles efficiently and stably integrate Cas9 and RFP linked by a 2A peptide and driven by the EF1 alpha promoter (EF1a-Cas9-2A-GFP) for strong expression in both dividing and non-dividing cell lines. They are ideal for avoiding the tedious lentivirus production process and are one part of a two part CRISPR system with individual Cas9 and gRNA expression vectors. For easy fluorescence microscopy a MOI greater than 200 should be employed.

To order gRNA in any format click here

Application

Functional Genomics/Target Validation
  • Creation of gene knockouts in multiple cell lines
  • Complete knockout of genes not amenable to RNAi
  • Creation of cell lines stably expressing Cas9-RFP

Features and Benefits

  • Highly specific
  • Highly active
  • Ready to infect/transduce

Principle

CRISPR/Cas systems are employed by bacteria and archaea as a defense against invading viruses and plasmids. Recently, the type II CRISPR/Cas system from the bacterium Streptococcus pyogenes has been engineered to function in eukaryotic systems using two molecular components: a single Cas9 protein and a non-coding guide RNA (gRNA). The Cas9 endonuclease can be programmed with a single gRNA, directing a DNA double-strand break (DSB) at a desired genomic location. Similar to DSBs induced by zinc finger nucleases (ZFNs), the cell then activates endogenous DNA repair processes, either non-homologous end joining (NHEJ) or homology-directed repair (HDR), to heal the targeted DSB.

Unit Definition

VP/ml is the concentration unit of measure for viral titer estimated by p24 assay.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service