Skip to Content
Merck
All Photos(1)

Key Documents

C9619

Sigma-Aldrich

CAMK1δ, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CKLiK, CaMKID, RP11-462F151.1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$537.00

$537.00


Estimated to ship on30 April 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$537.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$537.00


Estimated to ship on30 April 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

117-159 nmol/min·mg

mol wt

~68 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CAMK1D(57118)

Biochem/physiol Actions

CAMK1δ or Ca2+/calmodulin-dependent kinase I-like kinase (CKLiK) is activated by Ca2+ and calmodulin and is detected in CD34+-derived neutrophils and eosinophils, as well as in mature peripheral blood granulocytes.CAMK1δ exhibits Ca2+/CaM-dependent activity that is enhanced in vitro by phosphorylation of its Thr180 by CaM-K kinase (CaM-KK)alpha, consistent with detection of CAMK1δ-activating activity in HeLa cells.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Verploegen et al.
Blood, 106(3), 1076-1083 (2005-04-21)
Activation of granulocyte effector functions, such as induction of the respiratory burst and migration, are regulated by a variety of relatively ill-defined signaling pathways. Recently, we identified a novel Ca2+/calmodulin-dependent kinase I-like kinase, CKLiK, which exhibits restricted mRNA expression to
Yumi Ishikawa et al.
FEBS letters, 550(1-3), 57-63 (2003-08-26)
In this report, we cloned a novel calmodulin-kinase (CaM-KIdelta) from HeLa cells and characterized its activation mechanism. CaM-KIdelta exhibits Ca(2+)/CaM-dependent activity that is enhanced (approximately 30-fold) in vitro by phosphorylation of its Thr180 by CaM-K kinase (CaM-KK)alpha, consistent with detection

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service