Skip to Content
Merck
All Photos(1)

Key Documents

C9415

Sigma-Aldrich

Z-Arg-Arg-Pro-Phe-His-Sta-Ile-His-Nε-Boc-Lys methyl ester

≥85% (HPLC)

Synonym(s):

CGP-29287

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C72H110N20O15
CAS Number:
Molecular Weight:
1495.77
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥85% (HPLC)

form

solid

technique(s)

ligand binding assay: suitable

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)C[C@H](O)[C@H](CC(C)C)NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@@H]3CCCN3C(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)OCc4ccccc4)C(=O)N[C@@H](Cc5c[nH]cn5)C(=O)N[C@@H](CCCCNC(=O)OC(C)(C)C)C(=O)OC

InChI

1S/C72H110N20O15/c1-9-44(4)59(65(100)89-55(36-48-39-78-42-83-48)62(97)85-51(67(102)105-8)25-16-17-29-81-70(103)107-72(5,6)7)91-58(94)37-57(93)52(33-43(2)3)86-63(98)54(35-47-38-77-41-82-47)87-61(96)53(34-45-21-12-10-13-22-45)88-64(99)56-28-20-32-92(56)66(101)50(27-19-31-80-69(75)76)84-60(95)49(26-18-30-79-68(73)74)90-71(104)106-40-46-23-14-11-15-24-46/h10-15,21-24,38-39,41-44,49-57,59,93H,9,16-20,25-37,40H2,1-8H3,(H,77,82)(H,78,83)(H,81,103)(H,84,95)(H,85,97)(H,86,98)(H,87,96)(H,88,99)(H,89,100)(H,90,104)(H,91,94)(H4,73,74,79)(H4,75,76,80)/t44-,49-,50-,51-,52-,53-,54-,55-,56-,57-,59-/m0/s1

InChI key

DLAHCJHYEAZDLE-VRYQDWSQSA-N

Biochem/physiol Actions

Primate-specific renin inhibitor with prolonged duration of action.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Nussberger et al.
Life sciences, 42(17), 1683-1688 (1988-01-01)
To prevent in vitro generation of angiotensins, the renin inhibitor CGP 29287 (CGP) was added to blood sampling tubes. Plasma immunoreactive angiotensin (ir-ANG) I and II were simultaneously measured by radioimmunoassay after rapid and quantitative extraction from a single plasma
M M de Gasparo et al.
Hypertension (Dallas, Tex. : 1979), 11(3), 209-216 (1988-03-01)
The mechanism by which monoclonal antibodies directed against human renin (R3-36-16 and R3-47-10) inhibit renin activity was investigated using various substrates. Both antibodies acted as potent inhibitors of human renin activity when human angiotensinogen was used as a substrate. However
J M Wood et al.
Journal of hypertension. Supplement : official journal of the International Society of Hypertension, 7(2), S37-S42 (1989-04-01)
We have developed marmoset models for the in vivo evaluation of primate-specific inhibitors of human renin. After acute intravenous administration to normotensive sodium-depleted marmosets, renin inhibitors of different structural types induced a maximum hypotensive response of a magnitude similar to
J M Wood et al.
Journal of cardiovascular pharmacology, 10 Suppl 7, S96-S98 (1987-01-01)
In this study, the hypotensive efficacy of a renin inhibitor was investigated during chronic administration. The renin inhibitor CGP 29,287 was administered by continuous intraperitoneal infusion with osmotic mini-pumps to normotensive marmosets fed a low-sodium diet. Marmosets received a dose
J M Wood et al.
Clinical and experimental hypertension. Part A, Theory and practice, 9(2-3), 337-343 (1987-01-01)
The primate-specific renin inhibitor CGP 29287 (30 mg/kg/d, n = 5) or the converting-enzyme inhibitor CGS 14831 (30 mg/kg/d, n = 8) were administered by continuous intraperitoneal infusion via osmotic minipumps to normotensive marmosets fed a low salt diet. CGP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service