Skip to Content
Merck
All Photos(1)

Key Documents

C8604

Sigma-Aldrich

Anti-CCR-5, N-Terminal antibody produced in rabbit

1 mg/mL, IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CCR5(1234)

General description

CCR5 is a chemokine receptor belonging to the super family of seven-transmembrane G-protein coupled receptors that mediates cellular functions of immune response, leukocyte migration, development and angiogenesis. CCR5 is expressed on macrophages and Th1 cells activated on inflammation, osteoclasts and vascular smooth muscle cells. Three CC chemokines, CCL3, 4 and 5 bind efficiently to CCR5, whereas CCL7, 8 and 13 bind less efficiently. The expression of CCR5 protein is most important in resistance to HIV-1 and hepatitis C virus infections and reportedly influences the course and progression of the infection. Anti-CCR5, N-Terminal specifically recognizes N terminal of human CCR5 (C-C chemokine receptor type 5).

Immunogen

synthetic peptide corresponding to the N-terminal amino acids 6-20 of human CCR5.

Application

Anti-CCR5, N-Terminal antibody may be used for western blot at 1 μg/mL and for immunohistochemistry at 2.5 μg/mL.

Physical form

Solution in 0.01 M phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemokine receptors: keys to AIDS pathogenesis?
D R Littman
Cell, 93(5), 677-680 (1998-06-18)
How do viruses enter cells? The HIV coreceptors teach us a lesson of complexity.
D S Dimitrov
Cell, 91(6), 721-730 (1997-12-31)
Martin Coenen et al.
European journal of medical research, 15(3), 97-101 (2010-05-11)
Efficient recruitment and activation of immuno-competent cells is crucial for an effective immune response to hepatitis C virus (HCV) infection. Chemokines and chemokine receptors have been shown to be critically involved in these processes. The CCR5 chemokine receptor is expressed
Ghalib Alkhatib
Current opinion in HIV and AIDS, 4(2), 96-103 (2009-04-03)
We discuss the current knowledge concerning the biology of CXCR4 and CCR5 and their roles in HIV-1 infection. Important research findings reported in the last 2 years have advanced our knowledge in the field of HIV coreceptors and pathogenesis. Novel
K L Jones et al.
British journal of pharmacology, 162(7), 1453-1469 (2010-12-08)
There is increasing recognition of an important contribution of chemokines and their receptors in the pathology of atherosclerosis and related cardiovascular disease. The chemokine receptor CCR5 was initially known for its role as a co-receptor for HIV infection of macrophages

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service