Skip to Content
Merck
All Photos(1)

Documents

C8239

Sigma-Aldrich

Anti-α-N-Catenin antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 102 kDa

species reactivity

mouse

technique(s)

microarray: suitable
western blot: 1:1,000 using cytosolic fraction of mouse brain

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Ctnna2(12386)

General description

The catenins (α, β, γ) are cytoplasmic proteins found in varying developing and adult tissues. α- N-catenin is a cadherin-binding protein, that plays a crucial role in cadherin-mediated cell-cell adhesion. It is widely expressed in the nervous system. α-Catenin (CAP102, 102 kDa), originally described as an E-cadherin associated protein, has been shown to associate with N-cadherin and P-cadherin. Within its conserved region α-catenin shows 30% identity to vinculin. There are at least two subtypes of α-catenin: α-E-catenin and α-N-catenin (102 kDa). The predominant form is α-E-catenin. Alternative spliced forms of α-N-catenin include α-N-catenin I and α-N-catenin II. The ratio of the two isoforms changes during development: isoforms II is more abundant than isoform I in early embryonic development, whereas isoform I is predominant in the adult stage.

Immunogen

synthetic peptide corresponding to a region located near the N-terminus of human α-N-catenin (amino acids 171-186) conjugated to KLH. This sequence is identical in mouse and chicken α-N-catenin. It is not found in α-E-catenin, β-catenin, and γ-catenin.

Application

α-N-Catenin antibody is suitable for microarray and western blot at a dilution of 1:1,000 by using cytosolic fraction of mouse brain.

Biochem/physiol Actions

Catenins bind directly or indirectly to the conserved cytoplasmic tail domain of the cell adhesion cadherins. The association of catenins to cadherins produces a complex, which is linked to the actin filament network. Catenins/cadherin complexes play an important role in mediating cell adhesion, transduction of cell-cell contact positional signals to the cell interior, and may play a crucial role in cell differentiation. The expression of α-N-catenin is more restricted and this form predominates in the brain and is localized at synaptic junctions.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unraveling the cytoplasmic interactions of the cadherin superfamily.
P Cowin
Proceedings of the National Academy of Sciences of the United States of America, 91(23), 10759-10761 (1994-11-08)
K Herrenknecht et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(20), 9156-9160 (1991-10-15)
The cytoplasmic region of the Ca(2+)-dependent cell-adhesion molecule (CAM) uvomorulin associates with distinct cytoplasmic proteins with molecular masses of 102, 88, and 80 kDa termed alpha, beta, and gamma catenin, respectively. This complex formation links uvomorulin to the actin filament
Angel Céspedes-Rubio et al.
Journal of neuroscience research, 88(16), 3621-3634 (2010-10-12)
Atorvastatin (ATV), a 3-hydroxy 3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor, exerts beneficial effects on stroke through several pleiotropic mechanisms. However, its role following cerebral ischemia is not completely understood yet. We evaluated the effect of ATV treatment on the synaptic
S Hirano et al.
Cell, 70(2), 293-301 (1992-07-24)
The function of cadherin cell adhesion molecules is thought to be regulated by a group of cytoplasmic proteins, including alpha-catenin. We identified a subtype of alpha-catenin, termed alpha N-catenin, which is associated with N-cadherin and expressed mainly in the nervous
Masato Uemura et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 235(9), 2559-2566 (2006-05-13)
Alpha N-catenin is a cadherin-binding protein, widely expressed in the nervous system; and it plays a crucial role in cadherin-mediated cell-cell adhesion. Here we report the effects of alpha N-catenin gene deficiency on brain morphogenesis. In addition to the previously

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service