Skip to Content
Merck
All Photos(1)

Key Documents

C7624

Sigma-Aldrich

Collagen human

Bornstein and Traub Type I, recombinant, expressed in Nicotiana tabacum (tobacco)

Synonym(s):

Collage

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human

Quality Level

recombinant

expressed in Nicotiana tabacum (tobacco)

Assay

>90% (SDS-PAGE)

form

liquid

concentration

2.5-3.5 mg/mL protein (Sircol method)

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Do not confuse Sigma type designations with recognized collagen classification types.

Physical form

Supplied as a filter-sterilized liquid in 10 mM HCl. Do not freeze.

Preparation Note

Coating of tissue culture dishes: Use 3 to 10 microgram per 1 cm2 dish area. For each cell type and plate type an optimization may be required. Dilute in 10 mM HCl or PBS. Apply the diluted collagen to a tissue culture plate and allow to dry overnight in a laminar flow hood with the plate lid open. Aspirate gently the remaining liquid from the coated wells. Wash twice gently with PBS.
To prepare a collagen gel the fibrillogenesis buffer contains 162 mM sodium phosphate dibasic (Na2HPO4) adjusted to pH 11.2 with 10 N NaOH, filter sterilized. Mix 9 volumes of Collagen at a concentration of 0.3%-1% with 1 volume of fibrillogenesis buffer. Mix well and incubate for 4 to 16 hours at 25°C to 27 °C. Higher concentration of collagen solution might need optimization. Thiis preparation will not spontaneously form a gel in PBS or cell culture medium.
Preparation of collagen sponge: concentrate gel to 10 mg/ml - 100 mg/ml by centrifugation, freeze dry. Crosslink by common methods found in the scientific literature such as 1-Ethyl-3-[3-dimethylaminopropyl]carbodiimide hydrochloride (EDC), Glutaraldehyde or thermal dehydration.
Other forms: membranes, sheets, fibers can be prepared by methods commonly used with other types of collagen.

Legal Information

Collage is a trademark of CollPlant Ltd.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Collagen: The Fibrous Proteins of the Matrix
Lodish H, et al.
Molecular and Cellular Biology (2000)
Mahboobe Ghaedi et al.
The Journal of clinical investigation, 123(11), 4950-4962 (2013-10-19)
The use of induced pluripotent stem cells (iPSCs) has been postulated to be the most effective strategy for developing patient-specific respiratory epithelial cells, which may be valuable for lung-related cell therapy and lung tissue engineering. We generated a relatively homogeneous
Davide Gibellini et al.
Retrovirology, 8, 40-40 (2011-05-27)
HIV infection elicits the onset of a progressive immunodeficiency and also damages several other organs and tissues such as the CNS, kidney, heart, blood vessels, adipose tissue and bone. In particular, HIV infection has been related to an increased incidence
Anne Tristan et al.
PloS one, 4(4), e5042-e5042 (2009-04-07)
Staphylococcus aureus necrotizing pneumonia is a severe disease caused by S. aureus strains carrying the Panton Valentine leukocidin (PVL) genes (lukS-PV & lukF-PV) encoded on various bacteriophages (such as phiSLT). Clinical PVL+ strains isolated from necrotizing pneumonia display an increased
Yao Xiong et al.
Frontiers in microbiology, 11, 590435-590435 (2020-12-18)
Adhesion to the gastrointestinal tract is considered to be important for bifidobacteria to colonize the human gut and exert their probiotic effects. Some cell surface proteins of bifidobacteria, known as adhesins, play critical roles in the binding to host cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service