Skip to Content
Merck
All Photos(1)

Key Documents

C7214

Sigma-Aldrich

Monoclonal Anti-Cyclin D3 antibody produced in mouse

clone DCS-22, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

clone

DCS-22, monoclonal

mol wt

antigen 31-34 kDa

contains

15 mM sodium azide

species reactivity

canine, human, rat, monkey, mouse

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 1:4,000 using a rat osteosarcoma cell line (ROS) extract

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CCND3(896)
mouse ... Ccnd3(12445)
rat ... Ccnd3(25193)

Specificity

It does not cross-react with other D-type cyclins.

Immunogen

recombinant human cyclin D3 protein.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stacey A Leisenfelder et al.
Journal of virology, 80(11), 5577-5587 (2006-05-16)
In its course of human infection, varicella-zoster virus (VZV) infects rarely dividing cells such as dermal fibroblasts, differentiated keratinocytes, mature T cells, and neurons, none of which are actively synthesizing DNA; however, VZV is able to productively infect them and
J Bartkova et al.
International journal of cancer, 65(3), 323-327 (1996-01-26)
The D-type cyclins are positive regulators of the G1 phase of the mammalian cell cycle. Cyclins D1 or D2 are over-expressed in several types of cancer, transform rodent cells in culture and therefore harbor hallmarks of cellular proto-oncogenes. In contrast
Daxiang Cui et al.
Toxicology letters, 155(1), 73-85 (2004-12-09)
The influence of single-walled carbon nanotubes (SWCNTs) on human HEK293 cells is investigated with the aim of exploring SWCNTs biocompatibility. Results showed that SWCNTs can inhibit HEK293 cell proliferation, decrease cell adhesive ability in a dose- and time-dependent manner. HEK293
Sandra Herrero-González et al.
Glia, 57(2), 222-233 (2008-08-30)
In previous studies, we showed that endothelin-1 increased astrocyte proliferation and glucose uptake. These effects were similar to those observed with other gap junction inhibitors, such as carbenoxolone (CBX). Because 24-h treatment with endothelin-1 or CBX downregulates the expression of
Mary Tsikitis et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(34), 12129-12134 (2005-08-16)
Rhabdoid tumors are aggressive pediatric malignancies for which, currently, there are no effective or standard treatment strategies. Rhabdoid tumors arise because of the loss of the tumor suppressor gene INI1. We have previously demonstrated that INI1 represses Cyclin D1 transcription

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service