Skip to Content
Merck
All Photos(2)

Key Documents

C7041

Sigma-Aldrich

McN-A-343

≥98% (TLC)

Synonym(s):

(4-Hydroxy-2-butynyl)-1-trimethyl­ammonium-3-chloro­carbanilate chloride, 4-[N-(3-Chlorophenyl)carbamoyloxy]-2-­butynyl­trimethyl­ammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18Cl2N2O2
CAS Number:
Molecular Weight:
317.21
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

color

off-white

solubility

H2O: soluble
ethanol: soluble

SMILES string

[Cl-].C[N+](C)(C)CC#CCOC(=O)Nc1cccc(Cl)c1

InChI

1S/C14H17ClN2O2.ClH/c1-17(2,3)9-4-5-10-19-14(18)16-13-8-6-7-12(15)11-13;/h6-8,11H,9-10H2,1-3H3;1H

InChI key

CXFZFEJJLNLOTA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

M1 muscarinic acetylcholine receptor agonist.

Preparation Note

Solutions may be stored for several days at 4 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren T May et al.
Molecular pharmacology, 72(2), 463-476 (2007-05-26)
The M2 muscarinic acetylcholine receptor (mAChR) possesses at least one binding site for allosteric modulators that is dependent on the residues (172)EDGE(175), Tyr(177), and Thr(423). However, the contribution of these residues to actions of allosteric agonists, as opposed to modulators
Hinako Suga et al.
Molecular pharmacology, 78(4), 745-755 (2010-07-21)
Investigating how a test drug alters the reaction of a site-directed electrophile with a receptor is a powerful method for determining whether the drug acts competitively or allosterically, provided that the binding site of the electrophile is known. In this
Laura Oliveira et al.
Neuro-Signals, 14(5), 262-272 (2005-11-23)
At the rat motor endplate, pre-synaptic facilitatory adenosine A2A and muscarinic M1 receptors are mutually exclusive. We investigated whether these receptors share a common intracellular signalling pathway. Suppression of McN-A-343-induced M1 facilitation of [3H]ACh release was partially recovered when CGS21680C
Hinako Suga et al.
The Journal of pharmacology and experimental therapeutics, 327(2), 518-528 (2008-08-07)
We explored the interaction of a nitrogen mustard derivative of acetylcholine with the human M(2) muscarinic receptor expressed in Chinese hamster ovary cells using the muscarinic radioligand, [3H]N-methylscopolamine (NMS). Acetylcholine mustard caused a concentration-dependent, first-order loss of [3H]NMS binding at
Frederick J Mitchelson
Pharmacology & therapeutics, 135(2), 216-245 (2012-05-31)
The unusual pharmacology of McN-A-343 was first described by Roszowski in 1961. The agonist appeared to be a selective stimulant of muscarinic receptors in sympathetic ganglia, now known to be the muscarinic M₁ receptor subtype. However, subsequent research demonstrated that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service