Skip to Content
Merck
All Photos(1)

Key Documents

C6608

Sigma-Aldrich

N-Acetyl-Lys-Pro-Arg-7-amido-4-trifluoromethylcoumarin

≥95% (HPLC), solid

Synonym(s):

Ac-KPR-AFC

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$672.00

$672.00


Estimated to ship on30 May 2025



Select a Size

Change View
10 MG
$672.00

About This Item

Empirical Formula (Hill Notation):
C29H39N8O6F3
Molecular Weight:
652.67
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

$672.00


Estimated to ship on30 May 2025


Product Name

N-Acetyl-Lys-Pro-Arg-7-amido-4-trifluoromethylcoumarin, ≥95% (HPLC), solid

Quality Level

Assay

≥95% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H](CCCCN)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CCCNC(N)=N)C(=O)Nc2ccc3c(OC(=O)C=C3C(F)(F)F)c2

InChI

1S/C29H39F3N8O6/c1-16(41)37-21(6-2-3-11-33)27(45)40-13-5-8-22(40)26(44)39-20(7-4-12-36-28(34)35)25(43)38-17-9-10-18-19(29(30,31)32)15-24(42)46-23(18)14-17/h9-10,14-15,20-22H,2-8,11-13,33H2,1H3,(H,37,41)(H,38,43)(H,39,44)(H4,34,35,36)/t20-,21-,22-/m0/s1

InChI key

UVABJZWXZFWYGH-FKBYEOEOSA-N

Substrates

Tryptase substrate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabsang Lee et al.
Nature protocols, 5(4), 688-701 (2010-04-03)
Human pluripotent stem cell (hPSC)-derived neural crest (NC) cells present a valuable tool for modeling aspects of human NC development, including cell fate specification, multipotency and cell migration. hPSC-derived NC cells are also suitable for modeling human disease and as
Laura Menendez et al.
Nature protocols, 8(1), 203-212 (2013-01-05)
Multipotent neural crest stem cells (NCSCs) have the potential to generate a wide range of cell types including melanocytes; peripheral neurons; and smooth muscle, bone, cartilage and fat cells. This protocol describes in detail how to perform a highly efficient

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service