Skip to Content
Merck
All Photos(1)

Key Documents

C6206

Sigma-Aldrich

N-Carboxymethyl-6-(2,2-dicyanovinyl)-1,2,3,4-tetrahydroquinoline

≥98% purity (HPLC), powder

Synonym(s):

CDCQ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O2
CAS Number:
Molecular Weight:
267.28
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

N-Carboxymethyl-6-(2,2-dicyanovinyl)-1,2,3,4-tetrahydroquinoline, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: 17.5 mg/mL, clear, bright yellow

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

OC(=O)CN1CCCc2cc(ccc12)\C=C(\C#N)C#N

InChI

1S/C15H13N3O2/c16-8-12(9-17)6-11-3-4-14-13(7-11)2-1-5-18(14)10-15(19)20/h3-4,6-7H,1-2,5,10H2,(H,19,20)

InChI key

LSKCTKMTYNICJW-UHFFFAOYSA-N

Other Notes

Fluorescent molecular rotor

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Iio et al.
Journal of biochemistry, 113(2), 196-199 (1993-02-01)
1-(2-Hydroxyethyl)-6-[(2,2-dicyano)vinyl]-2,3,4-trihydroquinoli ne (DCQ) is a fluorescent dye and its fluorescence yield is determined by a freedom of intramolecular rotation. When DCQ was added to an actin solution, its fluorescence intensity increased as the actin molecule transformed from G-actin to F-actin.
T Iwaki et al.
Biochemistry, 32(29), 7589-7592 (1993-07-27)
We have prepared monoclonal antibodies for the fluorescent molecular rotors 9-(2-carboxy-2-cyanovinyl)julolidine (CCVJ) and 9-(dicyanovinyl)julolidine (DCVJ). Mouse monoclonal antibody (IgG2b) prepared against CCVJ-conjugated bovine serum albumin strongly bound CCVJ and DCVJ. The CCVJ (or DCVJ) binding to IgG and Fab was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service