Skip to Content
Merck
All Photos(1)

Key Documents

C5856

Sigma-Aldrich

Charybdotoxin, recombinant from Leiurus quinquestriatus hebraeus

recombinant, expressed in E. coli, lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
$205.00
10 μG
$320.00

$205.00


Estimated to ship on28 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 μG
$205.00
10 μG
$320.00

About This Item

Empirical Formula (Hill Notation):
C176H280N58O55S7
Molecular Weight:
4312.92
UNSPSC Code:
51111800
NACRES:
NA.77

$205.00


Estimated to ship on28 May 2025

New, lower price on this item!

Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

form

lyophilized powder

storage temp.

−20°C

Amino Acid Sequence

Gln-Phe-Thr-Asn-Val-Ser-Cys-Thr-Thr-Ser-Lys-Glu-Cys-Trp-Ser-Val-Cys-Gln-Arg-Leu-His-Asn-Thr-Ser-Arg-Gly-Lys-Cys-Met-Asn-Lys-Lys-Cys-Arg-Cys-Tyr-Ser

Biochem/physiol Actions

Exhibits the same activity as the natural form of the toxin

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E E Sugg et al.
The Journal of biological chemistry, 265(31), 18745-18748 (1990-11-05)
Charybdotoxin (ChTX), a potent inhibitor of the high conductance Ca2(+)-activated K+ channel (PK,Ca) is a highly basic peptide isolated from venom of the scorpion Leiurus quinquestriatus hebraeus, whose primary structure has been determined (Gimenez-Gallego, G., Navia, M. A., Reuben, J.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service