Skip to Content
Merck
All Photos(1)

Key Documents

C5820

Sigma-Aldrich

Chondroitin disaccharide Δdi-UA-2S sodium salt

Synonym(s):

α-ΔUA-2S-[1→3]-GalNAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H19NO14SNa2
CAS Number:
Molecular Weight:
503.34
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

synthetic

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

storage temp.

−20°C

SMILES string

[Na].CC(=O)NC(C=O)C(OC1OC(=CC(O)C1OS(O)(=O)=O)C(O)=O)C(O)C(O)CO

InChI

1S/C14H21NO14S.Na.H/c1-5(18)15-6(3-16)11(10(21)8(20)4-17)28-14-12(29-30(24,25)26)7(19)2-9(27-14)13(22)23;;/h2-3,6-8,10-12,14,17,19-21H,4H2,1H3,(H,15,18)(H,22,23)(H,24,25,26);;

InChI key

RZWFDDVYBLJQSX-UHFFFAOYSA-N

Application

Chondroitin, a glucosaminoglycan (GAG), is a polysaccharide chain of alternating units of N-acetylgalactosamine (GalNAc) and glucuronic acid (GlcA) that can be sulfated on the either or both GalNAc and GlcA units. Chondroitin and its sulfates are frequently attached to proteins to form proteoglycans.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

ΔUA = 4-deoxy-L-threo-hex-4-enopyranosyluronic acid; GalNAc = N-acetyl-D-galactosamine; 2S = 2-sulfate

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of chondroitin sulfate proteoglycans in regeneration and plasticity in the central nervous system.
Galtrey CM, Fawcett JW.
Brain Research. Brain Research Reviews, 54, 1-18 (2007)
J C F Kwok et al.
The international journal of biochemistry & cell biology, 44(4), 582-586 (2012-01-24)
Chondroitin sulfate is a glycosaminoglycan composed of N-acetylgalactosamine and glucuronic acid. It attaches to a core protein to form chondroitin sulfate proteoglycan (CSPG). Being a major component of the brain extracellular matrix, CSPGs are involved in neural development, axon pathfinding
Wen-Li Gu et al.
Sheng li ke xue jin zhan [Progress in physiology], 38(2), 101-105 (2007-07-18)
Chondroitin sulfate proteoglycans (CSPGs) are major components of extracellular matrix in the central nervous system (CNS) , and play important roles in the development and maintenance of CNS, as welt in pathogenesis or repair processes of neuronal damages. They mainly
Yulin Li et al.
Chemical Society reviews, 41(6), 2193-2221 (2011-11-26)
Injectable hydrogels with biodegradability have in situ formability which in vitro/in vivo allows an effective and homogeneous encapsulation of drugs/cells, and convenient in vivo surgical operation in a minimally invasive way, causing smaller scar size and less pain for patients.
Juliane Salbach et al.
Journal of molecular medicine (Berlin, Germany), 90(6), 625-635 (2011-12-22)
To meet the growing need for tissue replacement materials for our aging population, the development of new adaptive biomaterials is essential. The tissues with the highest demand for implant materials are skin and bone. These tissues share various similarities, including

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service