Skip to Content
Merck
All Photos(1)

Key Documents

C5740

Sigma-Aldrich

Cytochrome P450 human

2E1 isozyme microsomes, with P450 Reductase and cytochrome b5, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$790.00

$790.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
$790.00

About This Item

EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

$790.00


Check Cart for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected insect cells (BTI-TN-5B1-4)

description

1.0 nmole P450 2E1 per 0.5 ml vial

form

buffered aqueous solution (100 mM potassium phosphate, pH 7.4)

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CYP2E1(1571)

Looking for similar products? Visit Product Comparison Guide

Application

Cytochrome P450 is a heterogeneous family of isozymes whose primary function is to oxidize small molecules both as a function of intermediary metabolism (e.g., fatty acids) and to detoxify exogenous compounds (drugs or toxins). Alterations in the renal formation of Cytochrome P450 metabolites from arachidonic acid may participate in the formation of hypertension. Therefore, they have been identified as candidate mediators in the development of this multifactorial disease.

Biochem/physiol Actions

Cytochrome P450 is a heterogeneous family of isozymes whose primary function is to oxidize small molecules, both as a function of intermediary metabolism (e.g., fatty acids) and to detoxify exogenous compounds (drugs or toxins). Some isoforms have narrow substrate specificity, while others are promiscuous. The CYP1A1 isoform catalyzes 7-deethylation of ethoxyresorufin. Cytochrome P450 (CYP) plays an important role in detoxifying xenobiotics, cellular metabolism and homeostasis. One of the main mechanisms of drug-drug interactions is the induction or inhibition of these enzymes. CYP enzymes are transcriptionally activated by a variety of xenobiotics and by endogenous substrates via receptor-dependent pathways. Inhibition of these enzymes is a major factor in metabolism-based drug-drug interactions, and many chemotherapeutic medications can cause drug interactions by either inhibiting or inducing the cytochrome p450 enzyme system.

Analysis Note

Cytochrome c Reductase, b5, P450 content and p-Nitrophenol Hydroxylase activity reported on a lot-specific basis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mong-Heng Wang et al.
Hypertension (Dallas, Tex. : 1979), 42(4), 594-599 (2003-08-27)
The incidence of essential hypertension increases with obesity; however, the mechanisms that link obesity with hypertension are unclear. Renal cytochrome P450 (CYP)-derived eicosanoids--hydroxyeicosatetraenoic acids (HETEs), epoxyeicosatrienoic acids (EETs), and dihydroxyeicosatrienoic acids (DHETs)--have been shown to play an important role in
Xiangrong Zhang et al.
PloS one, 9(4), e94962-e94962 (2014-04-17)
The present study characterized in vitro metabolites of 20(R)-25-methoxyl-dammarane-3β, 12β, 20-triol (20(R)-25-OCH3-PPD) in mouse, rat, dog, monkey and human liver microsomes. 20(R)-25-OCH3-PPD was incubated with liver microsomes in the presence of NADPH. The reaction mixtures and the metabolites were identified
Mirza Bojić et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(9), 1438-1446 (2014-07-06)
Cilengitide is a stable cyclic pentapeptide containing an Arg-Gly-Asp motif responsible for selective binding to αVβ3 and αVβ5 integrins. The candidate drug showed unexpected inhibition of cytochrome P450 (P450) 3A4 at high concentrations, that is, a 15-mM concentration caused attenuation
Dongju Lin et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(10), 1727-1736 (2014-07-16)
Diosbulbin B (DIOB), a furan-containing diterpenoid lactone, is the most abundant component of Dioscorea bulbifera L. (DB), a traditional Chinese medicine herb. Administration of purified DIOB or DB extracts has been reported to cause liver injury in animals. The mechanisms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service