Skip to Content
Merck
All Photos(1)

Key Documents

C4547

Sigma-Aldrich

Cys-Gly-Tyr-Gly-Pro-Lys-Lys-Lys-Arg-Lys-Val-Gly-Gly

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H104N20O15S
CAS Number:
Molecular Weight:
1377.66
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥90% (HPLC)

composition

Peptide content, ~65%

storage temp.

−20°C

SMILES string

CC(C)C(NC(=O)C(CCCCN)NC(=O)C(CCCNC(N)=N)NC(=O)C(CCCCN)NC(=O)C(CCCCN)NC(=O)C(CCCCN)NC(=O)C1CCCN1C(=O)CNC(=O)C(Cc2ccc(O)cc2)NC(=O)CNC(=O)C(N)CS)C(=O)NCC(=O)NCC(O)=O

Biochem/physiol Actions

A SV40 T-antigen homolog capable of inducing nuclear transport

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R E Lanford et al.
Cell, 46(4), 575-582 (1986-08-15)
A system was developed for the analysis of protein transport to the nucleus. Carrier proteins cross-linked to synthetic peptides were microinjected into the cytoplasm of mammalian cells, and protein transport was evaluated by immunofluorescence staining of fixed cells. A 13-mer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service