Skip to Content
Merck
All Photos(1)

Key Documents

C4375

Sigma-Aldrich

N-Carbamyl-L-glutamic acid

≥98.0% (TLC)

Synonym(s):

N-(aminocarbonyl)-L-glutamic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$335.00

$335.00


Available to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
$335.00

About This Item

Empirical Formula (Hill Notation):
C6H10N2O5
CAS Number:
Molecular Weight:
190.15
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

$335.00


Available to ship on01 April 2025Details


Request a Bulk Order

Product Name

N-Carbamyl-L-glutamic acid,

Assay

≥98.0% (TLC)

form

powder

color

white

application(s)

cell analysis

storage temp.

2-8°C

SMILES string

NC(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C6H10N2O5/c7-6(13)8-3(5(11)12)1-2-4(9)10/h3H,1-2H2,(H,9,10)(H,11,12)(H3,7,8,13)/t3-/m0/s1

InChI key

LCQLHJZYVOQKHU-VKHMYHEASA-N

Gene Information

human ... CPS1(1373)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

N-Carbamyl-L-glutamic acid (carglumic acid) is use to study its potential as a replacement therapy for N-acetylglutamate synthetase (NAGS) deficiency and to treat hyperammonemia.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Gebhardt et al.
Journal of inherited metabolic disease, 28(2), 241-244 (2005-05-07)
In patients with propionic aciduria, the accumulating metabolite propionyl-CoA causes a disturbance of the urea cycle via the inhibition of N-acetylglutamate synthesis. Lack of this allosteric activator results in an inhibition of carbamoylphosphate synthase (CPS). This finally leads to hyperammonaemia.
Nicolas Gauthier et al.
PloS one, 8(7), e60581-e60581 (2013-07-19)
Most conditions detected by expanded newborn screening result from deficiency of one of the enzymes that degrade acyl-coenzyme A (CoA) esters in mitochondria. The role of acyl-CoAs in the pathophysiology of these disorders is poorly understood, in part because CoA
Xin Wu et al.
Amino acids, 39(3), 831-839 (2010-03-10)
This study determined effects of dietary supplementation with L-arginine (Arg) or N-carbamylglutamate (NCG) on intestinal health and growth in early-weaned pigs. Eighty-four Landrace x Yorkshire pigs (average body weight of 5.56+/-0.07 kg; weaned at 21 days of age) were fed
Luca Filippi et al.
Neonatology, 97(3), 286-290 (2009-11-06)
In propionic aciduria and methylmalonic aciduria, hyperammonemia as a symptom of metabolic decompensation is one of the major clinical problems. Hyperammonemia is a true neonatal emergency with high mortality and neurological complications in most survivors. It requires a rapid and
Ljubica Caldovic et al.
The Journal of pediatrics, 145(4), 552-554 (2004-10-14)
In a patient with N-acetylglutamate synthase (NAGS) deficiency, incorporation of an isotopic label from ammonium chloride into urea was markedly reduced before treatment with N-carbamyl-L-glutamate (NCLG) and completely normalized following treatment. Blood ammonia rose following ammonium tracer ingestion before treatment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service