Skip to Content
Merck
All Photos(1)

Key Documents

C-39110

Sigma-Aldrich

Mammary Epithelial Cell Growth Medium

SupplementPack containing all media supplements as individual vials, 1 Pack for 500 ml

Synonym(s):

Recommended with C-21210

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$346.00

$346.00


Check Cart for Availability


Select a Size

Change View
1 KIT
$346.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

$346.00


Check Cart for Availability

form

liquid

packaging

pkg of 1 kit

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Mammary Epithelial Cell Growth Medium SupplementPack (basal medium sold separately)
To form a complete medium, it should be combined with the basal medium: C-21210

Application

Mammary Epithelial Cell Growth Medium is a serum-free medium developed for the cultivation of epithelial cells from the mammary gland. The medium is optimized for primary human cells but can also be used for rat and murine mammary epithelial cells as well as mamma carcinoma cell lines.Recommended for:Human Mammary Epithelial Cells

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials

Quality

All lots of PromoCell Mammary Epithelial Cell Growth Medium are subjected to comprehensive quality control tests using primary human mammary epithelial cells. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested mammary epithelial cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Preparation Note

Final supplement concentrations after addition to the basal medium:Bovine Pituitary Extract: 0.004 ml / mlEpidermal Growth Factor (recombinant human): 10 ng / mlInsulin (recombinant human): 5 µg / mlHydrocortisone: 0.5 µg / ml

Storage and Stability

Store the SupplementPack at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Subculture Routine

Click here for more information.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dominika Czerwonka et al.
ChemMedChem, 15(2), 236-246 (2019-11-09)
The polyether ionophore salinomycin (SAL) has captured much interest because of its potent activity against cancer cells and cancer stem cells. Our previous studies have indicated that C1/C20 double-modification of SAL is a useful strategy to generate diverse agents with

Articles

Serum-free Mammary Epithelial Cell Growth Media optimized for the in vitro cultivation of epithelial cells from the mammary gland. Protocols for handling, cell passaging, media, and product use.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service