Skip to Content
Merck
All Photos(1)

Documents

C3484

Sigma-Aldrich

5-Carboxyrhodamine 6G, succinimidyl ester

Synonym(s):

5-CR 6G, SE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H29N3O7
Molecular Weight:
555.58
MDL number:
UNSPSC Code:
12171500

fluorescence

λex ~520 nm; λem ~550 nm

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

Fluorophore exhibiting excitation and emission spectra that fall between those of fluorescein and tetramethylrhodamine. Exhibits a higher fluorescence quantum yield than tetramethylrhodamine conjugates as well as excellent photostability.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service