Skip to Content
Merck
All Photos(1)

Key Documents

C-34006

Sigma-Aldrich

Calcium Chloride Solution

CaCl 2-Solution (0.5 M), 2 ml

Synonym(s):

Calcium Chloride Liquid

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$73.40

$73.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 ML
$73.40

About This Item

UNSPSC Code:
12352207
NACRES:
NA.81
grade:
for analytical purposes
form:
solution

$73.40


Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

form

solution

packaging

pkg of 2 ml

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

CaCl 2-Solution

Application

Some types of primary human cells, especially Normal Human Epidermal Keratinocytes (NHEK), respond sensitively to the calcium concentration in the culture medium. Calcium Chloride solution allows the adjustment of the optimal amount of calcium required by the cells. PromoCell Calcium Chloride (CaCl2) Solution is prepared as a solution from highly pure tissue culture tested Calcium Chloride and cell culture-grade water. It is a clear colorless and sterile solution.The concentration of CaCl2 is 0.5 M.

Quality

All lots of PromoCell Calcium Chloride Solution are subjected to comprehensive quality control tests. Each lot is routinely tested for the use with human primary cells, absence of cytotoxicity, and physical parameters such as osmolality and pH level. Approved in-house lots are used as a reference. In addition, all lots have been tested for the absence of microbial contaminants (fungi, bacteria).

Storage and Stability

Store at -20°C in the dark immediately after arrival. If stored properly, the product is stable until the expiry date stated on the label.

Subculture Routine

Click here for more information.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. I received this product for my master assay, and I saved it in -20 C. What should i do in order to use it and to change its concentration? Thank you so much for your time,

    1 answer
    1. Unfortunately, a protocol for dilution is not available. However, the following information from the product manual may be helpful. Some types of primary human cells, especially Normal Human Epidermal Keratinocytes, respond sensitively to the calcium concentration in the culture medium. Calcium Chloride solution allows the adjustment of the optimal amount of calcium required by the cells. The Calcium Chloride (CaCl2) Solution is prepared as a solution from highly pure tissue culture tested Calcium Chloride and cell culture-grade water. It is a clear, colorless, and sterile solution. The concentration of CaCl2 is 0.5 M, and each vial contains 2 mL.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service