Skip to Content
Merck
All Photos(1)

Key Documents

C3116

Sigma-Aldrich

Anti-CXCR4 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CXC Chemokine Receptor 4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
conjugate:
unconjugated
application:
IHC (p)
clone:
polyclonal
species reactivity:
human
citations:
7
technique(s):
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 9-14 μg/mL

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 9-14 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CXCR4(7852)
mouse ... Cxcr4(12767)
rat ... Cxcr4(60628)

Immunogen

synthetic peptide corresponding to the third cytoplasmic loop of human CXC chemokine receptor 4 (CXCR4), conjugated to KLH. The immunizing peptide has 100% homology with the rat and mouse gene.

Application

Anti-CXCR4 antibody produced in rabbit is suitable for immunohistochemistry of formalin-fixed, paraffin-embedded tissue sections at a working concentration of 9-14 μg/mL. It was used to stain rat bladder sections at a dilution of 1:2000, incubated overnight.

Biochem/physiol Actions

CXCR4 is a cell surface receptor of the chemokine CXCL12 (SDF-1) and is involved in metastasis, hematopoiesis and immune responses. It acts with CD4 to facilitate the entry of HIV into target cells. CXCR4 has important roles in tumor initiation, proliferation, invasion and metastasis of breast cancer cells.

Physical form

Solution in phosphate buffered saline, containing 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Walters et al.
British journal of cancer, 110(5), 1179-1188 (2014-01-16)
In experimental models of glioblastoma multiforme (GBM), irradiation (IR) induces local expression of the chemokine CXCL12/SDF-1, which promotes tumour recurrence. The role of CXCR7, the high-affinity receptor for CXCL12, in the tumour's response to IR has not been addressed. We
Angelo Ferrari et al.
BMC veterinary research, 8, 27-27 (2012-03-16)
Mammary tumours frequently develop in female domestic cats being highly malignant in a large percentage of cases. Chemokines regulate many physiological and pathological processes including organogenesis, chemotaxis of inflammatory cells, as well as tumour progression and metastasization. In particular, the
Elisabetta Razzuoli et al.
Veterinary research communications, 47(3), 1615-1627 (2023-04-11)
Canine Soft Tissue Sarcoma (STS) cell line A-72 has been largely employed for antiviral and antiproliferative studies. However, there are few information on their characteristics. Our aim was to evaluate A-72 expression level of genes and proteins involved in the
Bikash Debnath et al.
Theranostics, 3(1), 47-75 (2013-02-06)
CXCR4 is a G-protein-coupled receptor involved in a number of physiological processes in the hematopoietic and immune systems. The SDF-1/CXCR4 axis is significantly associated with several diseases, such as HIV, cancer, WHIM syndrome, rheumatoid arthritis, pulmonary fibrosis and lupus. For
Amnon Peled et al.
Theranostics, 3(1), 34-39 (2013-02-06)
The Chemokine receptor CXCR4 and its ligand stromal derived factor-1 (SDF-1/CXCL12) are important players involved in cross-talk between leukemia cells and the bone marrow (BM) microenvironment. CXCR4 expression is associated with poor prognosis in AML patients with and without the

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service